missing translation for 'onlineSavingsMsg'
Learn More

Erlab Socket 10/16A CH

Product Code. 31794116 Shop All Erlab Products
Change view
Click to view available options
For Use With (Equipment):
Laboratory Hood
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31794116 Laboratory Hood Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31794116 Supplier Erlab Supplier No. I2119000041
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The built-in power socket complements the design of the filtration fume cupboard by providing an accessible power supply point, tailored to the needs of laboratory applications.

  • Service Fittings for Laboratory Enclosures: These service fittings allow water, gas, compressed air and electrical or data connections to be integrated directly inside Erlab enclosures, so that operators have all the utilities they need at the point of use, without compromising containment.
  • Safe Integration Inside the Working Volume:
    • Components designed for installation in Erlab enclosure panels
    • Sealed, leak-tight assembly that preserves the containment of the working volume
    • Corrosion-resistant materials suitable for laboratory environments
    • Positioning studied to keep the working surface clear and ergonomic
    • Electrical accessories compliant with applicable safety requirements
  • Practical, Ready-to-Install Solutions: Supplied as a complete kit with the required sealing and fixing components, these fittings can be installed on new equipment or retrofitted to an existing enclosure according to Erlab instructions.
  • Key Benefits:
    • Utilities available directly at the point of use.
    • Leak-tight installation that preserves enclosure containment.
    • Corrosion-resistant, easy-to-clean materials.
    • Ready-to-install kit designed for Erlab equipment.
    • Improved operator comfort and workstation efficiency.

Specifications

For Use With (Equipment) Laboratory Hood
Product Type Socket
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.