missing translation for 'onlineSavingsMsg'
Learn More

Sobetirome, MedChemExpress

Product Code. 16468787
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468787 5 mg 5mg
16452327 50 mg 50mg
16406198 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16468787 Supplier Medchem Express Supplier No. HY14823
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sobetirome (GC-1) is a thyroid hormone receptor β (TRβ)-specific agonist which bind selectively to TRβ-1 with an EC50 of 0.16 μM.

Sobetirome (GC-1) is a thyroid hormone receptor β (TRβ)-specific agonist which bind selectively to TRβ-1 with an EC50 of 0.16 μM.

Specifications

Chemical Name or Material Sobetirome
CAS 211110-63-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H24O4
Quantity 5 mg
Synonym GC-1 QRX-431
Solubility Information DMSO : 100 mg/mL (304.51 mM; Need ultrasonic)
SMILES O=C(O)COC1=CC(C)=C(CC2=CC=C(O)C(C(C)C)=C2)C(C)=C1
Molecular Weight (g/mol) 328.4
Formula Weight 328.4
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.