missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ SOB Broth
Click to view available options
Broth Type:
SOB Broth
Unit Size:
500g
Specifications
Specifications
| Broth Type | SOB Broth |
| Format | Capsules |
| Product Type | Microbiology Media |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction