missing translation for 'onlineSavingsMsg'
Learn More

Melford SOB Broth, Capsules

Product Code. 30018806
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018806 1 kg 1kg
30018807 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018806 Supplier Melford Supplier No. S250401000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SOB Broth is recommended for use in cultivation of recombinant strains of E.

SOB Broth is recommended for use in cultivation of recombinant strains of E. Coli. It is a nutritionally rich growth medium for use in the preparation and transformation of competent cells.

Add two capsules to one liter of distilled water. Heat and stir gently to completely dissolve medium. Sterilize by autoclaving at 121°C for 15 minutes.

Final Solution contains:

Casein Enzymic Hydrolysate: 20 g/l

Yeast Extract: 5 g/l

Sodium Chloride: 0.5 g/l

Magnesium Sulfate: 2.4 g/l

Potassium Chloride: 0.186 g/l

Specifications

Media Type SOB Broth
Product Type Nutrient Broth
Quantity 1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.