missing translation for 'onlineSavingsMsg'
Learn More

Melford SOB Broth

Product Code. p-200086974
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 30018803

Brand: Melford S25000500.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SOB Broth is recommended for use in cultivation of recombinant strains of E.

SOB Broth is recommended for use in cultivation of recombinant strains of E. Coli. It is a nutritionally rich growth medium for use in the preparation and transformation of competent cells.

Add 28 grams of Broth in 1 liter of distilled water. Heat gently to dissolve the medium completely. Dispense in tubes and sterilize by autoclaving for 15 minutes

.

Ingredients:

Casein Enzymic Hydrolysate: 20 g/l

Magnesium Sulfate: 2.4 g/l

Yeast Extract: 5 g/l

Potassium Chloride: 0.186 g/l

Sodium Chloride: 0.5 g/l

Final pH(25°:C): 7.0+0.2

Specifications

Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.