missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOAT 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£174.00 - £255.00
Specifications
| Antigen | SOAT 2 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:100-1:500 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18619677
|
Novus Biologicals
NBP3-06319-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18640797
|
Novus Biologicals
NBP3-06319-50ul |
50 μg |
£174.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOAT 2 Polyclonal antibody specifically detects SOAT 2 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| SOAT 2 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ACACT2MGC116732, ACAT-2, ACAT2ARGP2, acyl Co-A: cholesterol acyltransferase 2, acyl coenzyme A:cholesterol acyltransferase 2, acyl-CoA:cholesterol acyltransferase 2, Acyl-coenzyme A:cholesterol acyltransferase 2, Cholesterol acyltransferase 2, EC 2.3.1.26, sterol O-acyltransferase 2 | |
| Produced in rabbits immunized with E. coli-derived Human SOAT 2 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.0) | |
| 8435 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title