missing translation for 'onlineSavingsMsg'
Learn More

SOAT 2 Antibody, DyLight 405, Novus Biologicals™

Product Code. 18376384 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18376384 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18376384 Supplier Novus Biologicals Supplier No. NBP306319V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SOAT 2 Polyclonal specifically detects SOAT 2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOAT 2
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 405
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ACACT2MGC116732, ACAT-2, ACAT2ARGP2, acyl Co-A: cholesterol acyltransferase 2, acyl coenzyme A:cholesterol acyltransferase 2, acyl-CoA:cholesterol acyltransferase 2, Acyl-coenzyme A:cholesterol acyltransferase 2, Cholesterol acyltransferase 2, EC 2.3.1.26, sterol O-acyltransferase 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SOAT 2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8435
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.