missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOAT 2 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06319AF488
This item is not returnable.
View return policy
Description
SOAT 2 Polyclonal specifically detects SOAT 2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| SOAT 2 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| ACACT2MGC116732, ACAT-2, ACAT2ARGP2, acyl Co-A: cholesterol acyltransferase 2, acyl coenzyme A:cholesterol acyltransferase 2, acyl-CoA:cholesterol acyltransferase 2, Acyl-coenzyme A:cholesterol acyltransferase 2, Cholesterol acyltransferase 2, EC 2.3.1.26, sterol O-acyltransferase 2 | |
| Produced in rabbits immunized with E. coli-derived Human SOAT 2 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 8435 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction