missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15721-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members, but does contain a SH3 domain near its N-terminus. This protein interacts with the cytoplasmic domains of the precursor but not the processed forms of a disintegrin and metalloprotease domain 9 and 15. This protein binds the beta-appendage domain of adaptor protein 2 and may function to assist adaptor protein 2 in its role at the plasma membrane. This protein interacts with activated Cdc42-associated kinase-2 to regulate the degradation of epidermal growth factor receptor protein.Specifications
| SNX9 | |
| Polyclonal | |
| Unconjugated | |
| SNX9 | |
| MST155, MSTP155, Protein SDP1, SDP1, SH3PX1, SH3PXD3A, SNX9, sorting nexin 9, WISP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51429, 66616, 683687 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q91VH2, Q9Y5X1 | |
| SNX9 | |
| SNX9 Fusion Protein Ag8440 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction