missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX9 Polyclonal Antibody

SNX9 Polyclonal Antibody
Brand: Thermo Scientific PA556734
This item is not returnable.
View return policy
Description
SNX9 Polyclonal Antibody for Western Blot, ICC/IF, IHC
Specifications
| SNX9 | |
| Polyclonal | |
| Unconjugated | |
| Snx9 | |
| 2700073N08Rik; MST155; MSTP155; OTTHUMP00000040083; Protein SDP1; RP11-266C7.10; SDP1; SH3 and PX domain-containing protein 1; SH3 and PX domain-containing protein 3A; SH3PX1; SH3PXD3A; Snx9; sorting nexin 9; sorting nexin-9; Wiskott-Aldrich syndrome protein (WASP) interactor protein; WISP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51429, 66616, 683687 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q91VH2, Q9Y5X1 | |
| Snx9 | |
| Recombinant protein corresponding to Human SNX9. Recombinant protein control fragment (Product #RP-94818). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction