missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNX8 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA563428

Product Code. 15761202

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: EKGVLHKHQR ALHKYSLMKR QMMSATAQNR EPESVEQLES RIVEQENAIQ TMELRNYFSL YCLHQETQLI HVYLPLTSHI LRAFVNSQIQ GHKEMSKVWN DLRPKLSCLF AGPHS Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

Sorting nexin (SNX) proteins are members of a large family of hydrophilic PX (phospholipid-binding motif) domain-containing proteins that interact with a variety of receptor types. SNXs are widely expressed, although the tissue distribution of each SNX mRNA varies. The ability of SNXs to bind specific phospholipids, as well as their tendency to form protein-protein complexes, suggests a role for these proteins in cellular membrane trafficking and protein sorting. SNXs may also function specifically in pro-degradative sorting, internalization, endosomal recycling or simply in endosomal sorting. SNXs partially associate with cellular membranes, despite their hydrophilic nature. SNX8 is an ortholog of a yeast protein.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SNX8
Polyclonal
Unconjugated
Snx8
B130023O14Rik; Mvp1; SNX8; sorting nexin 8; sorting nexin-8
Rabbit
Antigen affinity chromatography
RUO
29886
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9Y5X2
Snx8
Recombinant protein corresponding to Human SNX8. Recombinant protein control fragment (Product #RP-92668).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.