missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNX5 Recombinant Rabbit Monoclonal Antibody (HL2375)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550509
This item is not returnable.
View return policy
Description
SNX5 Recombinant Monoclonal Antibody for ICC/IF, IHC (P)
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein binds to fanconi anemia complementation group A protein, but its function is unknown. This gene results in two transcript variants encoding the same protein.
Specifications
| SNX5 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9D8U8, Q9Y5X3 | |
| SNX5 | |
| Recombinant fragment of human SNX5. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| HL2375 | |
| Unconjugated | |
| SNX5 | |
| 0910001N05Rik; 1810032P22Rik; AU019504; D2Ertd52e; FLJ10931; id:ibd5091; RP11-504H3.2; Snx5; snx5 protein; sorting nexin 5; sorting nexin-5; wu:fa66c10; wu:fi28h04; zgc:55769 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 27131, 69178 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction