missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNX5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550769
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total SNX5 protein.
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein binds to fanconi anemia complementation group A protein, but its function is unknown. This gene results in two transcript variants encoding the same protein.
Spécification
| SNX5 | |
| Polyclonal | |
| Unconjugated | |
| SNX5 | |
| 0910001N05Rik; 1810032P22Rik; AU019504; D2Ertd52e; FLJ10931; id:ibd5091; RP11-504H3.2; Snx5; snx5 protein; sorting nexin 5; sorting nexin-5; wu:fa66c10; wu:fi28h04; zgc:55769 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27131, 296199, 69178 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.6 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| B1H267, Q9D8U8, Q9Y5X3 | |
| SNX5 | |
| Synthetic peptide corresponding to a region derived from internal residues of human sorting nexin 5. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu