missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNX4 Recombinant Rabbit Monoclonal Antibody (K01_1Y62)

Product Code. 30826706
Change view
Click to view available options
Quantity:
50 μL
100 μL
Unit Size:
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30826706 50 μL 50µL
30826369 100 μL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826706 Supplier Invitrogen™ Supplier No. MA565078

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

SNX4 Recombinant Monoclonal Antibody for Western Blot

SNX2 and SNX4 share a high degree of amino acid similarity with SNX1, as they all contain a characteristic phox homology (PX) domain. These proteins are all partially associated with cellular membranes, and they, likewise, associate with EGF, PDGF and insulin receptor tyrosine kinases. These nexins are widely expressed and yet have various tissue distribution patterns. Additionally, the sorting nexins can associate with each other and with a variety of other cellular proteins, suggesting that they exist as part of multisubunit complexes. The related protein, SNX3, comprises a distinct subgroup of nexins that share less sequence similarity outside of the PX domain and have dramatically different binding affinities for the tyrosine kinase receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX4
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1Y62
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene SNX4
Gene Accession No. O95219, Q91YJ2
Gene Alias 1810036H14Rik; AI037066; ATG24B; Snx4; sorting nexin 4; sorting nexin-4
Gene Symbols SNX4
Host Species Rabbit
Immunogen A synthetic peptide of human SNX4.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360725, 69150, 8723
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.