missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX33 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20447
This item is not returnable.
View return policy
Description
SNX33 Polyclonal specifically detects SNX33 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SNX33 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| MGC32065, SH3 and PX domain containing 3, SH3PXD3CSH3PX3, SNX30SH3 and PX domain-containing protein 3, sorting nexin 33, sorting nexin-33 | |
| Rabbit | |
| 65 kDa | |
| 0.1 mL | |
| Signal Transduction | |
| 257364 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.91mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q8WV41 | |
| SNX33 | |
| Recombinant protein encompassing a sequence within the center region of human SNX33. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction