missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX32 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25763-1-AP-150UL
This item is not returnable.
View return policy
Description
SNX32 (sorting nexin-32), also known as SNX6B (sorting nexin-6B), is a 403 amino acid protein that contains one PX (phox homology) domain and belongs to the sorting nexin family. Existing as two alternatively spliced isoforms, SNX32 may be involved in several stages of intracellular trafficking.Specifications
| SNX32 | |
| Polyclonal | |
| Unconjugated | |
| SNX32 | |
| SNX32, SNX6B, sorting nexin 32, Sorting nexin 6B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 254122 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.46 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q86XE0 | |
| SNX32 | |
| SNX32 Fusion Protein Ag22653 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction