missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNX32 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596898
This item is not returnable.
View return policy
Description
Positive Samples: Mouse brain, Rat brain Immunogen sequence: HTRIRDACLR ADRVMRAHKC LADDYIPISA ALSSLGTQEV NQLRTSFLKL AELFERLRKL EGRVASDE.
SNX32 (sorting nexin-32), also known as SNX6B (sorting nexin-6B), is a 403 amino acid protein that contains one PX (phox homology) domain and belongs to the sorting nexin family. Existing as two alternatively spliced isoforms, SNX32 may be involved in several stages of intracellular trafficking.
Specifications
| SNX32 | |
| Polyclonal | |
| Unconjugated | |
| SNX32 | |
| B930037P14Rik; RGD1560591; SNX32; Snx6b; sortin nexin-6B; sorting nexin 32; sorting nexin 6B; sorting nexin-32; Sorting nexin-6B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 225861, 254122, 361708 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.81 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q80ZJ7, Q86XE0 | |
| SNX32 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 218-285 of human SNX32 (NP_6899732). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction