missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10772-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like most family members. This protein interacts with phosphatidylinositol-3-phosphate, and is involved in protein trafficking. A pseudogene of this gene is present on the sex chromosomes. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Specifications
| SNX3 | |
| Polyclonal | |
| Unconjugated | |
| SNX3 | |
| Grd19, MCOPS8, Protein SDP3, SDP3, SNX3, sorting nexin 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54198, 684097, 8724 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O60493, O70492, Q5U211 | |
| Snx3 | |
| SNX3 Fusion Protein Ag1160 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto