missing translation for 'onlineSavingsMsg'
Learn More

SNX3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA589203

Product Code. 16577181

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MAETVADTRR LITKPQNLND AYGPPSNFLE IDVSNPQTVG VGRGRFTTYE IRVKTNLPIF KLKESTVRRR YSDFEWLRSE LERESKVVVP PLPGKAFLRQ LPFRGDDGIF DDNFIEERKQ GLEQFINKVA GHPLAQNERC LHMFLQDEII DKSYTPSKIR HA; Positive Samples: HL-60, Mouse kidney, Mouse brain; Cellular Location: Cytoplasmic vesicle, Early endosome, phagosome

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like most family members. This protein interacts with phosphatidylinositol-3-phosphate, and is involved in protein trafficking. A pseudogene of this gene is present on the sex chromosomes. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SNX3
Polyclonal
Unconjugated
SNX3
Grd19, MCOPS8, Protein SDP3, SDP3, SNX 3A, Snx3, sorting nexin 3, sorting nexin 3A, sorting nexin-3
Rabbit
Affinity Chromatography
RUO
54198, 684097, 8724
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1.48 mg/mL
PBS with 50% glycerol and 0.02% sodium azide, pH 7.3
O60493, O70492, Q5U211
SNX3
Recombinant fusion protein containing a sequence corresponding to amino acids 1-162 of human SNX3 (NP_0037861)
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.