missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNX29 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552623
This item is not returnable.
View return policy
Description
Immunogen sequence: QGVSSLFREI TASSAVSILI KPEQETDPLP VVSRNVSADA KCKKERKKKK KVTNIISFDD EEDEQNSGDV FKKTPGAGES SEDNSDRSSV NIMSAFESPF GPNS Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 76%.
SNX29 also known as RUNDC2A, is a 375 amino protein that contains one RUN domain. RUNDC2A is encoded by a gene that maps to human chromosome 16p13.13, which encodes over 900 genes and comprises nearly 3% of the human genome. The GAN gene is located on chromosome 16 and, with mutation, may lead to giant axonal neuropathy, a nervous system disorder characterized by increasing malfunction with growth. The rare disorder Rubinstein-Taybi syndrome is also associated with chromosome 16, as is Crohn's disease, which is a gastrointestinal inflammatory condition.
Specifications
| SNX29 | |
| Polyclonal | |
| Unconjugated | |
| SNX29 | |
| 4933437K13Rik; A-388D4.1; ENSMUSG00000079738; Gm11170; Gm1737; Gm930; RUN domain containing 2A; RUN domain-containing protein 2A; RUNDC2A; SNX29; SODD; sorting nexin 29; sorting nexin-29 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 92017 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TEQ0 | |
| SNX29 | |
| Recombinant protein corresponding to Human SNX29. Recombinant protein control fragment (Product #RP-89452). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction