missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX27 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
POA - £407.00
Specifications
| Antigen | SNX27 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 ug/ml, Simple Western 10 ug/ml, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation reported in scientific literature (PMID 32413996), Immunohistochemistry-Paraffin 1:100 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18057695
|
Novus Biologicals
NBP1-45283SS |
0.025 mL |
POA
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18701044
|
Novus Biologicals
NBP1-45283 |
£407.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
SNX27 Polyclonal specifically detects SNX27 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| SNX27 | |
| Western Blot 1 ug/ml, Simple Western 10 ug/ml, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation reported in scientific literature (PMID 32413996), Immunohistochemistry-Paraffin 1:100 | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| KIAA0488MGC126873, methamphetamine-responsive transcript 1, MGC126871, MGC20471, MRT1, MY014, sorting nexin family member 27, sorting nexin-27 | |
| SNX27 | |
| IgG | |
| Affinity Purified | |
| Human and mouse. Predicted to react with rat, bovine and Zebrafish based on 100% sequence homology. |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| Q96L92, Q3UHD6 | |
| 81609 | |
| Synthetic peptide made to an internal portion of human SNX27 (within residues 450-500). [Swiss-Prot# Q96L92] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 65 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title