missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX26 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-53781
This item is not returnable.
View return policy
Description
SNX26 Polyclonal specifically detects SNX26 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| SNX26 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| FLJ39019, neurite outgrowth multiadaptor RhoGAP protein, NOMA-GAP, Rho GTPase activating protein 33, rho GTPase-activating protein 33, Rho-type GTPase-activating protein 33, sorting nexin 26, Sorting nexin-26, Tc10/CDC42 GTPase-activating protein, TCGAPSNX26 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 115703 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1.25 ug/ml, Immunohistochemistry-Paraffin 1:10 - 1:500, Peptide ELISA Detection limit 1:32000 | |
| NP_443180.2 | |
| ARHGAP33 | |
| Peptide with sequence ARSTDSLDGPGE-C corresponding to N-Terminus according to NP_443180.2. | |
| 0.1 mg | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog and Pig. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction