missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX22 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13062-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a sorting nexin that is found in the cytoplasm, where it interacts with membrane-bound phosphatidylinositol 3-phosphate. The encoded protein may play a role in intracellular trafficking. Two transcript variants, one protein-coding and the other not protein-coding, have been found for this gene.Specifications
| SNX22 | |
| Polyclonal | |
| Unconjugated | |
| SNX22 | |
| SNX22, sorting nexin 22 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 382083, 79856 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96L94 | |
| Snx22 | |
| SNX22 Fusion Protein Ag3751 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction