Learn More
SNX21 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22193-1-AP-20UL
 View more versions of this product
Product Code. 16851725
  • £135.00 / 20µL
Description
May be involved in several stages of intracellular trafficking.
Specifications
Specifications
SNX21
Polyclonal
Unconjugated
SNX21
C20orf161, PP3993, SNX L, SNX21, SNXL, Sorting nexin 21, sorting nexin family member 21, Sorting nexin L
Rabbit
Antigen Affinity Chromatography
RUO
90203
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.49 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q969T3
SNX21
SNX21 Fusion Protein Ag16900
20 μL