Learn More
Invitrogen™ SNX21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114205
Description
Immunogen sequence: DEDDEDEDDE EAGPDQLPLG DGTSGEDAER SPPPDGQWGS QLLARQLQDF For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 86%.
May be involved in several stages of intracellular trafficking.
Specifications
| SNX21 | |
| Polyclonal | |
| Unconjugated | |
| SNX21 | |
| C20orf161; dJ337O18.4; PP3993; SNX21; SNXL; SNX-L; sorting nexin 21; sorting nexin family member 21; sorting nexin L; Sorting nexin-21 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 90203 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q969T3 | |
| SNX21 | |
| Immunogen sequence: DEDDEDEDDE EAGPDQLPLG DGTSGEDAER SPPPDGQWGS QLLARQLQDF (aa 69-118). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.