missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£89.00 - £282.00
Specifications
| Antigen | SNX2 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18614574
|
Novus Biologicals
NBP3-21430-25ul |
25 μg |
£89.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663654
|
Novus Biologicals
NBP3-21430-100ul |
100 μg |
£282.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SNX2 Polyclonal antibody specifically detects SNX2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| SNX2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS, pH 7.0 | |
| 6643 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| MGC5204, sorting nexin 2, sorting nexin-2, Transformation-related gene 9 protein, TRG-9 | |
| Produced in rabbits immunized with E. coli-derived Human SNX2 fragment | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title