missing translation for 'onlineSavingsMsg'
Learn More

SNX2 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18431628 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431628 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431628 Supplier Novus Biologicals Supplier No. NBP224514AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SNX2 Polyclonal antibody specifically detects SNX2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX2
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias MGC5204, sorting nexin 2, sorting nexin-2, Transformation-related gene 9 protein, TRG-9
Host Species Rabbit
Immunogen A portion of amino acids 120-170 of human Snx2 was used as the immunogen.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6643
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.