missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX18 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21946-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members, but contains a SH3 domain. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| SNX18 | |
| Polyclonal | |
| Unconjugated | |
| SNX18 | |
| SH3PX2, SH3PXD3B, SNAG1, SNX18, sorting nexin 18 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 112574 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96RF0 | |
| SNX18 | |
| SNX18 Fusion Protein Ag16645 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction