missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNX15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583444
This item is not returnable.
View return policy
Description
Immunogen sequence: EEDGEGGPTP AYLSQATELI TQALRDEKAG AYAAALQGYR DGVHVLLQGV PSDPLPARQE GVKKKAAEYL KRAEEILRLH LSQLPP Highest antigen sequence indentity to the following orthologs: Mouse - 74%, Rat - 72%.
This gene encodes a member of the sorting nexin family. Members of this family contain a phox domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. Overexpression of this gene results in a decrease in the processing of insulin and hepatocyte growth factor receptors to their mature subunits. This decrease is caused by the mislocalization of furin, the endoprotease responsible for cleavage of insulin and hepatocyte growth factor receptors. This protein is involved in endosomal trafficking from the plasma membrane to recycling endosomes or the trans-Golgi network. Alternative splicing results in multiple transcript variants. Read-through transcription also exists between this gene and the upstream ADP-ribosylation factor-like 2 gene.
Specifications
| SNX15 | |
| Polyclonal | |
| Unconjugated | |
| Snx15 | |
| 1500032B08Rik; clone iota unknown protein; E130013C21Rik; hCG_23393; HSAF001435; OTTHUMP00000035515; SNX15; sorting nexin 15; sorting nexin-15 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29907 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NRS6 | |
| Snx15 | |
| Recombinant protein corresponding to Human SNX15. Recombinant protein control fragment (Product #RP-97287). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction