missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNX12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521774
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, H1299, HeLaS3. Predicted reactivity: Mouse (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members. A similar protein in mouse may be involved in regulating the neurite outgrowth. Alternate splicing results in multiple transcript variants.
Specifications
| SNX12 | |
| Polyclonal | |
| Unconjugated | |
| SNX12 | |
| 2610001F05Rik; AW045757; fc02c04; fc03c06; fj97e07; LOW QUALITY PROTEIN: sorting nexin-12; MGC118982; MGC118983; RGD1565585; SDP8; SDP8 protein; similar to sorting nexin 12; snx12; sorting nexin 12; sorting nexin 12, variation 1; sorting nexin 12, variation 2; sorting nexin-12; wu:fc02c04; wu:fc03c06; wu:fj97e07; zgc:158590; zgc:55728; zgc:77034; zgc:77034 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29934 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q9UMY4 | |
| SNX12 | |
| Synthetic peptide contain a sequence corresponding to a region within amino acids 100 and 112 () of SNX12. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction