missing translation for 'onlineSavingsMsg'
Learn More

SNX11 Antibody (2G1), Novus Biologicals™

Product Code. 18358169 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18358169 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18358169 Supplier Novus Biologicals Supplier No. H00029916M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SNX11 Monoclonal antibody specifically detects SNX11 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX11
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 2G1
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH00768
Gene Alias MGC111019, sorting nexin 11, sorting nexin-11
Host Species Mouse
Immunogen SNX11 (AAH00768, 1 a.a. ~ 270 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGFWCRMSENQEQEEVITVRVQDPRVQNEGSWNSYVDYKIFLHTNSKAFTAKTSCVRRRYREFVWLRKQLQRNAGLVPVPELPGKSTFFGTSDEFIEKRRQGLQHFLEKVLQSVVLLSDSQLHLFLQSQLSVPEIEACVQGRSTMTVSDAILRYAMSNCGWAQEERQSSSHLAKGDQPKSCCFLPRSGRRSSPSPPPSEEKDHLEVWAPVVDSEVPSLESPTLPPLSSPLCCDFGRPKEGTSTLQSVRRAVGGDHAVPLDPGQLETVLEK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 29916
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.