missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNX1 Polyclonal Antibody, Alexa Fluor™ 555

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA1836A555
This item is not returnable.
View return policy
Description
The PA1-836 immunogen is a synthetic peptide corresponding to residues Q(139) E D Q F D L T V G I T D P E K I(155) of human SNX1. This peptide (Cat. # PEP-286) is available for use in neutralization and control experiments.
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This endosomal protein regulates the cell-surface expression of epidermal growth factor receptor. This protein also has a role in sorting protease-activated receptor-1 from early endosomes to lysosomes. This protein may form oligomeric complexes with family members. This gene results in three transcript variants encoding distinct isoforms.
Specifications
| SNX1 | |
| Polyclonal | |
| Alexa Fluor 555 | |
| SNX1 | |
| HsT17379; MGC8664; Snx1; SNX1A; sorting nexin 1; sorting nexin 1A; sorting nexin-1; VPS5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56440, 6642 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Immunocytochemistry | |
| 1.0 mg/mL | |
| PBS with 1.0mg/mL BSA and 0.05% sodium azide | |
| Q13596, Q9WV80 | |
| SNX1 | |
| Synthetic peptide corresponding to residues Q(139) E D Q F D L T V G I T D P E K I(155) of human SNX1. | |
| 50 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido