missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SNW1 Antibody (PCRP-SNW1-1C12), Novus Biologicals™
Mouse Monoclonal Antibody
£178.00 - £351.00
Specifications
| Antigen | SNW1 |
|---|---|
| Clone | PCRP-SNW1-1C12 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30044008
|
Novus Biologicals™
NBP3-26781-20ug |
20 μg |
£178.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043600
|
Novus Biologicals™
NBP3-26781-100ug |
100 μg |
£351.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SNW1 Monoclonal antibody specifically detects SNW1 in Human samples. It is validated for Flow CytometrySpecifications
| SNW1 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Bx42, homolog of Drosophila BX42, MGC119379, NCOA-62, Nuclear protein SkiP, Nuclear receptor coactivator NCoA-62, nuclear receptor coactivator, 62-kD, Prp45, PRPF45, SKI interacting protein, Ski-interacting protein, SKIPSKIIP, SNW domain containing 1, SNW domain-containing protein 1 | |
| Recombinant fragment of human SNW1 protein | |
| Primary | |
| Store at 4°C. |
| PCRP-SNW1-1C12 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 22938 | |
| IgG2a | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title