missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNW1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35445-20ul
This item is not returnable.
View return policy
Description
SNW1 Polyclonal antibody specifically detects SNW1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| SNW1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:100 | |
| Bx42, homolog of Drosophila BX42, MGC119379, NCOA-62, Nuclear protein SkiP, Nuclear receptor coactivator NCoA-62, nuclear receptor coactivator, 62-kD, Prp45, PRPF45, SKI interacting protein, Ski-interacting protein, SKIPSKIIP, SNW domain containing 1, SNW domain-containing protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 300-536 of human SNW1 (NP_036377.1).,, Sequence:, EAVEMRAQVERKMAQKEKEKHEEKLREMAQKARERRAGIKTHVEKEDGEARERDEIRHDRRKERQHDRNLSRAAPDKRSKLQRNENRDISEVIALGVPNPRTSNEVQYDQRLFNQSKGMDSGFAGGEDEIYNVYDQAWRGGKDMAQSIYRPSKNLDKDMYGDDLEARIKTNRFVPDKEFSGSDRRQRGREGPVQFEEDPFGLDKFLEEAKQHGGSKRPSDSSRPKEHEHEGKKRRKE | |
| 20 μL | |
| Signal Transduction | |
| 22938 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction