missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNURPORTIN1 Antibody (SNUPN/7363R), Novus Biologicals™
Rabbit Monoclonal Antibody
£224.00 - £412.00
Specifications
| Antigen | SNURPORTIN1 |
|---|---|
| Clone | SNUPN/7363R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18623565
|
Novus Biologicals
NBP3-20383-100ug |
100 μg |
£412.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600305
|
Novus Biologicals
NBP3-20383-20ug |
20 μg |
£224.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
SNURPORTIN1 Monoclonal antibody specifically detects SNURPORTIN1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifikationer
| SNURPORTIN1 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| KPNBL, RNA U transporter 1, RNA, U transporter 1, RNUT1, snurportin 1, Snurportin1, SNURPORTIN-1, SPN1 | |
| Recombinant fragment (around aa 900-1100) of human SNURPORTIN1 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
| SNUPN/7363R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10mM PBS with 0.05% BSA | |
| 10073 | |
| IgG κ | |
| Protein A or G purified |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel