missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNTG2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA578505

Product Code. 16307204

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive Control: A549, H1299 Predicted Reactivity: Mouse (86%), Rat (86%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Syntrophin gamma 2 encodes a protein belonging to the syntrophin family. Syntrophins are cytoplasmic peripheral membrane proteins that bind to components of mechanosenstive sodium channels and the extreme carboxy-terminal domain of dystrophin and dystrophin-related proteins. The PDZ domain of this protein product interacts with a protein component of a mechanosensitive sodium channel that affects channel gating. Absence or reduction of this protein product has been associated with Duchenne muscular dystrophy. There is evidence of alternative splicing yet the full-length nature of these variants has not been described.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SNTG2
Polyclonal
Unconjugated
SNTG2
2210008K22Rik; 9530013L23; BB121248; G2SYN; Gamma-2-syntrophin; gamma2-syntrophin; MGC133174; SNTG2; SYN5; syntrophin; syntrophin 5; syntrophin gamma 2; syntrophin, gamma 2; syntrophin-5
Rabbit
Antigen Affinity Chromatography
RUO
54221
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 20% glycerol and 0.01% thimerosal; pH 7
Q9NY99
SNTG2
Recombinant protein encompassing a sequence within the center region of human Syntrophin gamma 2. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.