missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNTG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570303
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 86%.
Adapter protein that binds to and probably organizes the subcellular localization of a variety of proteins. May link various receptors to the actin cytoskeleton and the dystrophin glycoprotein complex. May participate in regulating the subcellular location of diacylglycerol kinase-zeta to ensure that diacylglycerol is rapidly inactivated following receptor activation.
Specifications
| SNTG1 | |
| Polyclonal | |
| Unconjugated | |
| SNTG1 | |
| 4933426D16Rik; G1SYN; Gamma-1-syntrophin; gamma1-syntrophin; RGD1560290; Sntg1; SYN4; syntrophin 4; syntrophin gamma 1; syntrophin, gamma 1; syntrophin-4; zmp:0000001117 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 54212 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NSN8 | |
| SNTG1 | |
| synthetic peptide directed towards the middle region of human SNTG1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction