missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNTB2, Mouse anti-Human, Clone: 3F12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00006645-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: VRVVKQEAGGLGISIKGGRENRMPILISKIFPGLAADQSRALRLGDAILSVNGTDLRQATHDQAVQALKRAGKEVLLEVKFIREVTPYIKKPSLVSpecifications
| SNTB2 | |
| Monoclonal | |
| Unconjugated | |
| syntrophin, beta 2 (dystrophin-associated protein A1, 59kDa, basic component 2) | |
| D16S2531E/EST25263/SNT2B2/SNT3/SNTL | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 3F12 | |
| In 1x PBS, pH 7.4 | |
| NM_006750 | |
| SNTB2 | |
| SNTB2 (NP_006741.1, 116 a.a. ∼ 210 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6645 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction