missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNTB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107273
This item is not returnable.
View return policy
Description
Sequence homology: Dog: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Adapter protein that binds to and probably organizes the subcellular localization of a variety of membrane proteins. May link various receptors to the actin cytoskeleton and the dystrophin glycoprotein complex.
Specifications
| SNTB1 | |
| Polyclonal | |
| Unconjugated | |
| SNTB1 | |
| 59 kDa dystrophin-associated protein A1 basic component 1; 59-1 DAP; 59-DAP; A1B; beta-1-syntrophin; beta1-Syntrophin; BSYN2; DAPA1B; dystrophin-associated protein A1, 59kD, basic component 1; SNT2; SNT2B1; SNTB1; syntrophin beta 1; syntrophin, basic 1; syntrophin, beta 1; syntrophin, beta 1 (dystrophin-associated protein A1, 59kDa, basic component 1); syntrophin-2; Tax interaction protein 43; TIP-43 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6641 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q13884 | |
| SNTB1 | |
| Synthetic peptide directed towards the N terminal region of human SNTB1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction