missing translation for 'onlineSavingsMsg'
Learn More

SNS-314 mesylate, MedChemExpress

Product Code. p-200099232
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16467657

Brand: Medchem Express HY12003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SNS-314 mesylate is a potent and selective aurora kinase inhibitor with IC50s of 9, 31, and 6 nM for aurora A, B and C, respectively.

SNS-314 mesylate is a potent and selective aurora kinase inhibitor with IC50s of 9, 31, and 6 nM for aurora A, B and C, respectively.

Specifications

Chemical Name or Material SNS-314 mesylate
CAS 1146618-41-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H19ClN6O4S3
Quantity 5 mg
Solubility Information DMSO : 150 mg/mL (284.61 mM; Need ultrasonic)
SMILES O=C(NC1=CC=CC(Cl)=C1)NC2=NC=C(S2)CCNC3=C4C(C=CS4)=NC=N3.OS(C)(=O)=O
Molecular Weight (g/mol) 527.04
Formula Weight 527.04
Percent Purity 99.53%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.