missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SNS 032
GREENER_CHOICE

Product Code. 18004252
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18004252

Brand: Tocris Bioscience™ 4075/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cdk inhibitor; potently inhibits cdk2, cdk7 and cdk9

Cyclin dependent kinase (cdk) inhibitor (reported IC50 values are 0.004 - 0.048, 0.0045 - 0.05, 0.062 - 0.29, 0.2-0.48, 0.4 and 0.45 μM for cdk9, cdk2, cdk7, cdk1, cdk5 and cdk4, respectively). Displays no activity against 190 additional kinases (IC50 >1 μM). Arrests the cell cycle at G2/M; inhibits transcription, proliferation and colony formation, and induces apoptosis in RPMI-8226 multiple myeloma cells. Prevents tumor cell-induced VEGF secretion and in vitro angiogenesis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 345627-80-7
Molecular Formula C17H24N4O2S2

Specifications

Quantity 50 mg
Formula Weight 380.53
Purity >99%
Molecular Weight (g/mol) 380.525
Chemical Name or Material SNS 032
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.