missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRPN Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82347-0.1ML
This item is not returnable.
View return policy
Description
SNRPN Polyclonal specifically detects SNRPN in Human samples. It is validated for Western Blot.
Specifications
| SNRPN | |
| Polyclonal | |
| PBS, 2% sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| DKFZp686C0927, DKFZp686M12165, DKFZp761I1912, DKFZp762N022, FLJ39265, HCERN3FLJ33569, MGC29886, Prader-Willi syndrome chromosome region, PWCR, RT-LI, Sm protein D, Sm protein N, small nuclear ribonucleoprotein polypeptide N, small nuclear ribonucleoprotein-associated protein N, Sm-D, sm-N, SmN, SMNFLJ36996, SNRNP-N, SNURF-SNRPN, tissue-specific splicing protein, Tissue-specific-splicing protein | |
| The immunogen is a synthetic peptide directed towards the middle region of human SNRPN | |
| 0.1 mL | |
| Diabetes Research | |
| 6638 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction