missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNRPB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551941
This item is not returnable.
View return policy
Description
Immunogen sequence: TFKAFDKHMN LILCDCDEFR KIKPKNAKQP EREEKRVLGL VLLRGENLVS MTVEGPPPKD TGIARVPLAG AAGGPGVGRA AGRGVPAGVP IPQ Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The protein encoded by this gene is one of several nuclear proteins that are found in common among U1, U2, U4/U6, and U5 small ribonucleoprotein particles (snRNPs). These snRNPs are involved in pre-mRNA splicing, and the encoded protein may also play a role in pre-mRNA splicing or snRNP structure. Autoantibodies from patients with systemic lupus erythematosus frequently recognize epitopes on the encoded protein. Two transcript variants encoding different isoforms (B and B') have been found for this gene.
Specifications
| SNRPB | |
| Polyclonal | |
| Unconjugated | |
| SNRPB | |
| AL024368; AU018828; B polypeptide of Sm protein; CCMS; COD; sm protein B; Sm protein B/B'; SM11; small nuclear ribonucleoprotein B; small nuclear ribonucleoprotein polypeptide B; small nuclear ribonucleoprotein polypeptides B and B'; small nuclear ribonucleoprotein polypeptides B and B1; small nuclear ribonucleoprotein-associated protein B; Small nuclear ribonucleoprotein-associated proteins B and B'; smB; sm-B; Sm-B/B'; SmB/B'; sm-B/Sm-B'; SmB/SmB'; SNRNP-B; Snrpb; SNRPB1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 171365, 20638, 6628 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P14678, P27048 | |
| SNRPB | |
| Recombinant protein corresponding to Human SNRPN. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction