missing translation for 'onlineSavingsMsg'
Learn More

SNRPN Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18433335 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433335 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433335 Supplier Novus Biologicals Supplier No. NBP281851AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SNRPN Polyclonal antibody specifically detects SNRPN in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRPN
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias DKFZp686C0927, DKFZp686M12165, DKFZp761I1912, DKFZp762N022, FLJ39265, HCERN3FLJ33569, MGC29886, Prader-Willi syndrome chromosome region, PWCR, RT-LI, Sm protein D, Sm protein N, small nuclear ribonucleoprotein polypeptide N, small nuclear ribonucleoprotein-associated protein N, Sm-D, sm-N, SmN, SMNFLJ36996, SNRNP-N, SNURF-SNRPN, tissue-specific splicing protein, Tissue-specific-splicing protein
Host Species Rabbit
Immunogen SNRPN antibody was raised against a 20 amino acid peptide near the amino terminus of human SNRPN. The immunogen is located within amino acids 30 - 80 of SNRPN. Amino Acid Squence: DEFRKIKPKNAKQPEREEK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research
Primary or Secondary Primary
Gene ID (Entrez) 6638
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.