Learn More
Invitrogen™ SNRPF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113696
Beskrivning
Immunogen sequence: GYMNMQLANT EEYIDGALSG HLGEVLIRCN NVLYIRGVEE EEEDGEMRE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Goat: 90%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 100%; Zebrafish: 100%.
SNRPF belongs to the snRNP Sm proteins family and is associated with snRNP U1, U2, U4/U6 and U5.
Specifikationer
| SNRPF | |
| Polyclonal | |
| Unconjugated | |
| SNRPF | |
| 2010013O18Rik; LOC615240 protein; PBSCF; Sm protein F; Small nuclear ribonucleoprotein F; small nuclear ribonucleoprotein polypeptide F; SMF; sm-F; snRNP-F; snrpf; snrpfl | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6636 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P62306 | |
| SNRPF | |
| Synthetic peptide directed towards the middle region of human SNRPF. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.