missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRPF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-86813
This item is not returnable.
View return policy
Description
SNRPF Polyclonal specifically detects SNRPF in Human samples. It is validated for Western Blot.
Specifications
| SNRPF | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| PBSCF, Sm protein F, small nuclear ribonucleoprotein polypeptide F, SMF, Sm-Fsmall nuclear ribonucleoprotein F, snRNP-F | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human SNRPF | |
| 100 μL | |
| metabolism | |
| 6636 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction