missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNRPE Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598165
This item is not returnable.
View return policy
Description
SNRPE Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
The protein encoded by SNRPE is a core component of U small nuclear ribonucleoproteins, which are key components of the pre-mRNA processing spliceosome. The encoded protein plays a role in the 3' end processing of histone transcripts. This protein is one of the targets in the autoimmune disease systemic lupus erythematosus, and mutations in SNRPE have been associated with hypotrichosis. Several pseudogenes of SNRPE have been identified.
Specifications
| SNRPE | |
| Polyclonal | |
| Unconjugated | |
| SNRPE | |
| AL022645; B raf; Braf; B-raf; C76690; HYPT11; sm protein E; small nuclear ribonucleoprotein E; small nuclear ribonucleoprotein polypeptide E; smE; Sm-E; snRNPE; snRNP-E; SNRPE | |
| Rabbit | |
| Protein G | |
| RUO | |
| 6635 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2.6 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P62304 | |
| SNRPE | |
| Recombinant Human Small nuclear ribonucleoprotein E protein (1-92AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction