missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNRPE Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578173
This item is not returnable.
View return policy
Description
Positive Control: 293T, HeLa, HepG2 Predicted Reactivity: Mouse (100%), Rat (100%), Pig (100%), Chicken (100%), Rhesus Monkey (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by SNRPE is a core component of U small nuclear ribonucleoproteins, which are key components of the pre-mRNA processing spliceosome. The encoded protein plays a role in the 3' end processing of histone transcripts. This protein is one of the targets in the autoimmune disease systemic lupus erythematosus, and mutations in SNRPE have been associated with hypotrichosis. Several pseudogenes of SNRPE have been identified.
Specifications
| SNRPE | |
| Polyclonal | |
| Unconjugated | |
| SNRPE | |
| AL022645; B raf; Braf; B-raf; C76690; HYPT11; sm protein E; small nuclear ribonucleoprotein E; small nuclear ribonucleoprotein polypeptide E; smE; Sm-E; snRNPE; snRNP-E; SNRPE | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20643, 6635 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.01 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P62304, P62305 | |
| SNRPE | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human SNRPE. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction