missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRPD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10379-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein belongs to the small nuclear ribonucleoprotein core protein family. It is required for pre-mRNA splicing and small nuclear ribonucleoprotein biogenesis.Specifications
| SNRPD3 | |
| Polyclonal | |
| Unconjugated | |
| SNRPD3 | |
| Sm D3, SMD3, snRNP core protein D3, SNRPD3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6634, 67332, 687711 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P62318, P62320 | |
| SNRPD3 | |
| SNRPD3 Fusion Protein Ag0293 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction