Learn More
Invitrogen™ SNRPD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527547
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (100%), Xenopus laevis (98%), Dog (100%), Pig (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene belongs to the small nuclear ribonucleoprotein core protein family. It is required for pre-mRNA splicing and small nuclear ribonucleoprotein biogenesis. Alternative splicing occurs at this locus and two transcript variants encoding the same protein have been identified.
Specifications
| SNRPD2 | |
| Polyclonal | |
| Unconjugated | |
| SNRPD2 | |
| 1810009A06Rik; small nuclear ribonucleoprotein D2; small nuclear ribonucleoprotein D2 polypeptide; small nuclear ribonucleoprotein D2 polypeptide 16.5kDa; small nuclear ribonucleoprotein Sm D2; SMD2; sm-D2; snRNP core protein D2; SNRPD1; SNRPD2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 107686, 6633 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.46 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P62316, P62317 | |
| SNRPD2 | |
| Recombinant protein encompassing a sequence within the center region of human SNRPD2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.